CryoCube® F740, Freezers, Eppendorf
Supplier: Eppendorf
CryoCube®
F740410035 F740210015 F740410015 F740420015 F740420035
76040-412EA 25474.2 USD
76040-412 76040-126 76040-410 76040-124 76040-406
CryoCube® F740, Freezers, Eppendorf
Freezers Ultra Low Temperature Freezers
The CryoCube® F740 ULT freezer combines the high-quality tradition of previous freezers with an increased capacity.
  • Easy to read, eye level, flush mounted control panel and alarm status
  • Reduces user stress
  • Supports an ergonomic workspace and epGreen
  • Provides optimal workflow in the lab
  • Automatic vent port, sealing inner doors
  • Advanced interface, electronic lock
The F740 series is designed to store more samples securely while maintaining optimal energy usage. Special low-temperature outer door seals allow gaskets to remain flexible at –86 °C and prevent ice buildup.
Traditionally foamed polyurethane insulation reinforced with vacuum insulation panels in freezer body and door (i versions) provide efficient insulation protection. Temperature and event logging, voltage stabilizer, 120 - 208 V switching. Full stainless steel interior for easy cleaning during maintenance and defrost cycles; powder-coated steel exterior.
CryoCube F740h: 80 mm wall (polyurethane foam and vacuum insulation panels), LED interface, green cooling liquids (R290/ R170), and air-cooling.
CryoCube F740hi: 80 mm wall (polyurethane foam and vacuum insulation panels), touchscreen interface, green cooling liquids (R290/ R170), and air-cooling.
Suitable for storage and preservation of all types of biological materials/samples.