INO-LOOP™ Inoculating Loops and Needles, Simport Scientific
Supplier: Simport
INO-LOOP™
L200-11 L200-21 L200-1 L200-3 L200-2A L200-2 L200-1A
10118-340CS 183.4 USD
10118-340 10118-332 10118-330 10118-342 10118-336 10118-334 10118-338
INO-LOOP™ Inoculating Loops and Needles, Simport Scientific
Spreaders Inoculating Loops and Needles
Smooth loop surface provides uniform and gentle streaking.
  • Do not require flaming
  • Sterile
  • Packed in safe, tamper-proof, zip-lock resealable bags
  • Made of high impact polystyrene
Loops and needles are smooth and flexible to facilitate uniform streaking without damaging the gel surface. Needles are straight and suitable for removal of specimens of single colonies. Packed sterile in safe, tamper-proof, zip-lock resealable bags.
Disposable inoculating loops and needles do not require flaming, and therefore eliminate the risk of infection due to aerosol formation of pathogenic substances.
Cross contamination due to improper sterilization is eliminated. They can be used under hoods without danger, and are color-coded for ease of size identification.
Ordering information: Certificate of conformity available upon request.