Safety Funnels, with Level Control, S.C.A.T
Supplier: SCAT Europe
117637
75972-506EA 582.96 USD
75972-506
Safety Funnels, with Level Control, S.C.A.T
Made of electrostatic conductive HDPE. Space saving construction, suitable for every safety cabinet.
  • Lightweight construction
  • Integrated level control
  • No more overflowing containers
  • Grounding cable and clamp included
  • Removable sieve for easy cleaning
Keep filling levels in non-transparent containers safe and easy under control.