Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Digital Camera, MC170 HD, Leica Microsystems
Supplier: Leica Microsystems
12730465
10037-578EA 2561.35 USD InStock
10037-578
Digital Camera, MC170 HD, Leica Microsystems
Cameras Digital Cameras
HD Digital microscope camera with c-mount interface that provides a full high definition live image with up to 30fps and a standard capture resolution of 5MP.
  • Standalone operation
  • Wireless remote control
  • Fast live and high quality digital images
  • Powerful Leica software
  • Two Year Warranty
The camera can be used standalone without PC or connected via USB cable. In standalone mode all camera commands can be made by a wireless remote control which allows you to comfortably set a white balance setting, capture images, show the gallery of captured images, display overlays onto the live or captured image and more.
In PC mode the camera offers full compatibility with LAS and its many add-on modules like interactive measurement or extended focus.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty