Doxycycline ELISA Kit, BioVision, Inc.

Supplier: BioVision
AB285232-96TEST
76305-666EA 1598.33 USD
76305-666
Doxycycline ELISA Kit, BioVision, Inc.
Assays ELISAs
A competitive ELISA kit for the quantitative measurement of doxycycline in tissue, honey, milk, serum.

  • Detection Range: 0.2 to 5.4 ppb
  • Sensitivity: <0.2 ppb
  • Detection limitation: 2 ppb for serum, 5 ppb tissue, honey and milk, 10 ppb for milk
  • Cross reaction: Doxycycline 100%, tetracycline, minocycline 100%, chlortetracycline 180%, oxytetracyline 40%
  • Easy, convenient and time-saving

Doxycycline is A synthetic derivative of oxytetracycline. Acts as a broad spectrum antibiotic and matrix metallo-proteinases (MMP) inhibitor.

Useful in studies involving wound healing and tissue remodeling. Common side effects include diarrhea, nausea, vomiting, a red rash, and an increased risk of a sunburn. Use during pregnancy or in young children may result in permanent problems with the teeth including changes in their color.

Standard techniques/instruments (HPLC or GC-MS) are utilized to detect doxycycline. However, these techniques are complex, expensive, laborious, and time-consuming. Immunoassay techniques, such as ELISAs are commonly preferred as a simple, reliable and rapid method for the quantification of doxycycline in various samples.

Doxycycline ELISA Kit is a competitive-based ELISA that can be used for the determination of this antibiotic in tissue, honey, milk and serum.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR