Femtotips®, Injection Capillaries, Sterile, Eppendorf
Supplier: Eppendorf
Femtotip®
930000035 930000035 930000043 930000043
89009-312PK 326.8 USD
CA53512-811 89009-312 CA47727-018 47727-018
Femtotips®, Injection Capillaries, Sterile, Eppendorf
Microinjection Capillaries
Features screw-in thread for quick assembly into FemtoJet® capillary holder. Individual testing of every Femtotips® using a flow parameter.
Caution: For research use only.