8-Bromo-cAMP sodium salt 10mM in DMSO, EZSolution™, sterile-filtered

Synonyms: 8-Br-cAMP8-Bromo-cAMP8- Bromoadenosine- 3', 5'-monophosphate8- Bromoadenosine- 3', 5'- cyclic monophosphate8-Br-Camp sodium salt

B2257-5
76303-486EA 93.39 USD
76303-486
8-Bromo-cAMP sodium salt 10mM in DMSO, EZSolution™, sterile-filtered
8-Bromo-cAMP sodium salt

A 10 mM (5 mg in 1163 μl), sterile-filtered solution of the cAMP analog, 8-Br-cAMP (Cat. No. 1837) in anhydrous DMSO.


  • A cell-permeable cAMP analog

Formula: C₁₀H₁₀BrN₅NaO₆P
MW: 430.08 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00005844
CAS Number: 76939-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR