3,4-Dichloro-1-benzothiophene-2-carbohydrazide

Warning

Synonyms: OGG1 inhibitor O83,4-Dichloro-benzo[b]thiophene-2-carboxylic acid hydrazideOGG1-IN-083,4-Dichlorobenzo[b]thiophene-2-carbohydrazide

B2235-5 B2235-25
76303-442EA 577.91 USD
76303-442 76304-282
3,4-Dichloro-1-benzothiophene-2-carbohydrazide
3,4-Dichloro-1-benzothiophene-2-carbohydrazide

OGG1 Inhibitor O8 is a potent inhibitor of 8-Oxoguanine DNA Glycosylase-1 (OGG1).


  • A potent 8-Oxoguanine DNA Glycosylase-1 (OGG1) inhibitor

Formula: C₉H₆Cl₂N₂OS
MW: 261.13 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01993639
CAS Number: 350997-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR