1-(2-Thiazolylazo)-2-naphthol
Synonyms: TAN1-(2-Thiazoylazo)-2-naphtholNSC-139021
B2199-500 B2199-100
76303-322EA 297.72 USD
76303-322 76303-320
1-(2-Thiazolylazo)-2-naphthol
1-(2-Thiazolylazo)-2-naphthol
NSC-139021 (ERGi-USU) is a highly selective inhibitor for the growth of ERG-positive cancer cells with IC₅₀s ranging from 30 to 400 nM. NSC-139021 displays minimal effect on normal prostate or endothelial cells or ERG-negative tumor cell lines.
  • A highly selective inhibitor for the growth of ERG-positive cancer cells