Megestrol acetate

Warning

Synonyms: 17alpha-Hydroxy-6-methyl-4,6-pregnadiene-3,20-dione 17-Acetate17alpha-Acethoxy-6-methyl-4,6-pregnadiene-3,20-dioneMegestrol 17-Acetate

B2193-500 B2193-100
76303-544EA 297.72 USD
76303-544 76303-304
Megestrol acetate
Megestrol acetate

Megestrol acetate is a synthetic derivative of the naturally occurring female sex hormone progesterone, with progestogenic, antiestrogenic, and antineoplastic activities. Also acts as an appetite stimulant.


  • A synthetic derivative of the naturally occurring female sex hormone progesterone

Formula: C₂₄H₃₂O₄
MW: 384.52 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00056470
CAS Number: 595-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR