Pibrentasvir ≥98%

Danger

Synonyms: ABT-530Dimethyl ((2S,2'S,3R,3'R)-((2S,2'S)-(((2R,5R)-1-(3,5-difluoro-4-(4-(4-fluorophenyl)piperidin-1-yl)phenyl)pyrrolidine-2,5-diyl)bis(5-fluoro-1H-benzo[d]imidazole-6,2-diyl))bis(pyrrolidine-2,1-diyl))bis(3-methoxy-1-oxobutane-1,2-diyl))dicarbamateDimethyl ((2S,2'S,3R,3'R)-((2S,2'S)-(((2R,5R)-1-(3,5-difluoro-4-(4-(4-fluorophenyl)piperidin-1-yl)phenyl)pyrrolidine-2,5-diyl)bis(6-fluoro-1H-benzo[d]imidazole-5,2-diyl))bis(pyrrolidine-2,1-diyl))bis(3-methoxy-1-oxobutane-1,2-diyl))dicarbamate

B2168-25 B2168-5
76303-204EA 110.91 USD
76303-204 76303-206
Pibrentasvir ≥98%
Pibrentasvir

Pibrentasvir is a direct acting antiviral agent and Hepatitis C virus (HCV) NS5A inhibitor that targets the viral RNA replication and virion assembly.


  • A direct acting antiviral agent and Hepatitis C virus (HCV) NS5A inhibitor

Formula: C₅₇H₆₅F₅N₁₀O₈
MW: 1113.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD30747902
CAS Number: 1353900-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR