Spectinomycin solution 100 mg/mL, sterile

Supplier: TEKNOVA INC

Synonyms: Salmosan TTrobicin

S9525 S9525
101454-196EA 158.41 USD
101454-196 CA101454-196
Spectinomycin solution 100 mg/mL, sterile
Spectinomycin

Spectinomycin is an antibiotic. Resistance to spectinomycin is used as a selectable marker in genetic engineering.

Formula: C₁₄H₂₄N₂O₇
Storage Temperature: Freezer
MDL Number: MFCD04041129
CAS Number: 1695-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR