LigaTrap Human IgA Purification Kit, ImmunoReagents
LT-146KIT LT-146
76276-258EA 327 USD
76276-258 76276-256
LigaTrap Human IgA Purification Kit, ImmunoReagents
LigaTrap® Human IgA purification kit with LT technology utilizes novel, patented ligands, designed for IgA antibody purification. Kit provides all the necessary reagents for fast, convenient micro-scale purification of human IgA in just ten easy steps.
  • Designed to purify human IgA
  • For in vitro laboratory use
  • Binding capacity for monoclonal human IgA is >15 mg/ml resin
  • Storage: 2 to 8 °C
LigaTrap® Human IgA purification resin is engineered to purify high quality human IgA antibodies from recombinant and monoclonal sources. Resins bind an exceptionally broad range of recombinant, monoclonal and human/animal source polyclonal immunoglobulins.
Each spin column can be used, regenerated, and used up to ten times with minimal
loss in binding capacity.
Delivery information: Each kit contains ten microspin columns prefilled with 0.10 ml of LigaTrap® Human IgA Purification Resin, buffers, and collection tubes.
Caution: Not for diagnostic or therapeutic use. Not for human or animal consumption.