Moticam® 1080BMH Microscope Camera, Motic Instruments
Supplier: Motic
1100600100861
76163-538EA 2160.68 USD
76163-538
Moticam® 1080BMH Microscope Camera, Motic Instruments
As an affordable multi-taksing microscopy camera, the Moticam 1080 is in a class of its own. This camera does not require a computer for standard operation as the user can make image adjustments and capture straight through the cameras on-board software. Capture up to 8 MP images directly onto an SD card or connect an external monitor through the true output and this Moticam 1080 allows you to view details as you have not seen before. Should you wish to use a computer at the same time, simply connect the camera through the USB port and enjoy simultaneous image transmission.
  • Software included: On-board computer-free image processing software, Motic Images Plus for PC, OSX and Linux
  • Output Possibilities: Simultaneous HDMI, USB, SD card
  • Twain and direct show driver suppoet device
If you are looking for an all-in-one solution, the 1080 BMH comes with an attached high-resolution screen boasting native 1920×1080 pixel resolution on just a 10" space allowing for crisp, clear and true colour images. Used in an educations, industrial or clinical area, the Moticam 1080 line is the jack of all trades in our Moticam lineup.
Delivery information: It includes HDMI cable, USB 2.0 cable (a male to a male cable), calibration slide, universal power supply, motic images plus 3.0 for PC/OSX/Linux.