Sodium formate ≥99.0% ACS, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Formic acid sodium salt
247596-500G
BJ247596-500GEA
126.5
USD
BJ247596-500G
Sodium formate ≥99.0% ACS, Fluka™
Sodium formate
Formula:
HCO₂Na MW: 68.01 g/mol Melting Pt: 255 °C Density: 1.92 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013101 CAS Number: 141-53-7 EINECS: 205-488-0 |
Specification Test Results
Identity (IR) | complying |
Assay | Min. 99.0 % |
Water insoluble matter | Max. 0.005 % |
Calcium (Ca) | Max. 0.005 % |
Iron (Fe) | Max. 5 ppm |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 0.001 % |
Sulfate (SO₄) | Max. 0.001 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...