2-Methoxy-1-naphthoic acid 98%
|
Warning
|
H60434-03
H60434-03
AAH60434-03EA
120.23
USD
CAAAH60434-03
AAH60434-03
2-Methoxy-1-naphthoic acid 98%
2-Methoxy-1-naphthoic acid
Formula:
C₁₂H₁₀O₃ Melting Pt: 177…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021532 EINECS: 213-429-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...