LT200 Protective Eyewear, HexArmor
Supplier: Hexarmor
11-16004-02
76207-814EA 20.47 USD
76207-814
LT200 Protective Eyewear, HexArmor
Safety Glasses
Safety eyewear should never be treated as one size fits all. Simply wearing any generic pair of safety eyewear isn’t enough to protect the eyes.
  • Panoramic lens with integrated side protectors and brow guard for total coverage and protection
  • Contoured nose bridge for comfort and fit
  • Dual-injection soft and hard components ensure a secure, pressure-free fit
  • Adaptive, flexible side arms with multiple inclinations to fit most face shapes
  • Side arms fold flat for easy storage
  • LTX™ Lens Technology for premium optical clarity
  • 100% UV protection
The LT200 with permanent anti-fog coatings. Featuring innovative adaptive technology, a wide panoramic lens with side protection, and soft flexible side arms ensuring maximum wearer acceptance.
Although it may decrease the possibility of injury, poorly or incorrectly fitting eyewear will put users at a higher risk for injury. HexArmor’s German-engineered dual-injection mold technology, allows the combination of soft and hard components into flex frames, making them comfortable and durable, yet highly adaptive.
They fit to most face widths or shapes, making wearability, protection, and compliance an easier endeavor.
Certifications: Meets Standards ANSI Z87.1.2015, CE EN 166, CSA 94.3.