ɑ-Cyano-4-hydroxycinnamic acid

Warning

Synonyms: α-CHCA2-Cyano-3-(4-hydroxyphenyl)acrylic Acidα-Cyano-4-hydroxycinnamic AcidCHCA

B2162-500 B2162-1000
76303-182EA 87.57 USD
76303-182 76303-180
ɑ-Cyano-4-hydroxycinnamic acid
alpha-Cyano-4-hydroxycinnamic acid

α-Cyano-4-hydroxycinnamate (4-CIN) is a monocarboxylate transporter (MCT) inhibitor. Displays antitumor and antiangiogenic activity in gliomas in vivo; decreases glycolytic metabolism, migration, and invasion in U251 cells. Also blocks lactate efflux from glioma cells and sensitizes cells to irradiation.


  • A monocarboxylate transporter (MCT) inhibitor

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00004204
CAS Number: 28166-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR