Bleomycin sulfate (mixture) (from Streptomyces verticillus), white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
0219030610
76204-124EA
902.45
USD
76204-124
Bleomycin sulfate (mixture) (from Streptomyces verticillus), white powder
Bleomycin sulfate (mixture)
Bleomycin Sulfate is an atineoplastic antibiotic. A mixture of cytotonic glycopeptide antibiotics which reacts with DNA and causes strand scission; they have also been shown to have a type of oxygen transferase activity. Bleomycin A2 is the main component of the bleomycin employed clinically. Bleomycins differ from one another in the terminal amine and show varying biological activity.
- Concentration : 10 - 100 µg/mL
- Key Applications : It is used in research as antibiotics, antifungal
- Application Areas : Chemicals|Life Sciences
- Product Type : Biochemicals
- Biochemical Category : pharmaceuticals
- Solubility : Very soluble in water, methanol; slightly soluble in ethanol. Practically insoluble in acetone, ethyl acetate, butyl acetate, ether.
Formula:
C₅₅H₈₄N₁₇O₂₁S₃·xH₂SO₄ MW: 1512.6 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00070310 CAS Number: 9041-93-4 |
Specification Test Results
Identity Test | Passes |
Source | Streptomyces verticillus |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...