TransferTip® Microinjection Capillaries, Eppendorf
Supplier: Eppendorf
5195000079
490015-792EA 887.95 USD
490015-792
TransferTip® Microinjection Capillaries, Eppendorf
Microinjection Capillaries
Microinjection capillaries for the transfer of ES cells into mouse blastocyst or morula.
  • 15 µm inner diameter
  • With 20° tip angle
  • Rigid flange, 1 mm length
  • Heat-formed spike eases penetration
  • Sterilized via validated gamma irradiation
  • Capillary safe for optimum protection