3M™ Scotch-Weld™ EPX Mixing Nozzle
Supplier: 3M
7000000952
76240-670CS 666.68 USD
76240-670
3M™ Scotch-Weld™ EPX Mixing Nozzle
Adhesives
Scotch-Weld™ EPX mixing nozzle is ideal for easy uniform application.
  • Provides thorough mixing and precise placement of two-component structural adhesives
  • Tapered tip can be cut for bead size
  • Available in a variety of designs to fit all cartridge sizes
  • Minimize adhesive waste
  • Easy to replace with new nozzle to use remaining adhesive in a cartridge
  • Used nozzle can be left on cartridge to provide a good seal until ready to continue dispensing
They mixes and applies two-part adhesives and potting compounds. And, the tapered tips can be cut for precise bead size.