The BVX filter unit is a two operator, extraction system used adjacent to the workbenches.
- Two station design for dynamic fume extraction
- New brushless motor, quiet operation
- Complete portability, placed under or next to the bench-top
- Audible alarm informs operators when filters need replacing
- Easy access on/off remote switch
Fitted with 2×50 mm (2") Ø arms or 1×63 mm (2,5") Ø arm. Main filter has a HEPA efficiency of 99.97% at 0.3 micron, and an activated carbon filter to remove additional gases.
Certifications: UL, CSA,CE Certified.
Delivery information: BVX-201-KIT contains BVX-201 unit, set of pre-HEPA, gas filters, remote power switch, set of 2 BVX-ARML flexible esd safe arm, 760 mm (30) long with arm clip. BVX-203 contains BVX-200 unit, set of pre-filters (FG-BVX200, pack of 5), gas filter (FG-BVX200 deep bed gas filter), remote power switch, with no arm included.