Ammonium hydrogen carbonate 99.0-101.0% (by acidimetry), EMPROVE® ESSENTIAL Ph. Eur., BP, E503
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Ammonium bicarbonate
1.01131.5000
1.01131.9029
EM1.01131.9029EA
1693
USD
CA1.01131.5000
EM1.01131.9029
Ammonium hydrogen carbonate 99.0-101.0% (by acidimetry), EMPROVE® ESSENTIAL Ph. Eur., BP, E503
Ammonium hydrogen carbonate
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated .
Conforms to the purity criteria on food additives according to the current European Commission Regulation.
Formula:
NH₄HCO₃ MW: 79.06 g/mol Melting Pt: 106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012138 CAS Number: 1066-33-7 EINECS: 213-911-5 |
Specification Test Results
Assay (Acidimetric) | 99.0 - 101.0% |
Identity | Passes Test |
pH-Value (5 %; Water) | 7.5 - 8.1 |
Chloride (Cl) | ≤0.001% |
Sulfate (SO₄) | ≤0.0030% |
Heavy Metals (as Pb) | ≤0.001% |
As (Arsenic) | ≤0.0002% |
Fe (Iron) | ≤0.0005% |
Hg (Mercury) | ≤0.0001% |
Pb (Lead) | ≤0.0002% |
Non-Volatile Matter | ≤0.0500% |
Residual Solvents (ICH Q3C) | Excluded by Manufacturing Process |
Corresponds to Ph. Eur., BP, E503 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...