Bonkote™ Pens and Replacement Tips, O.C. White
Supplier: O.C. White
BR-102 BON-102
76244-428PK 325.97 USD
76244-428 76246-006
Bonkote™ Pens and Replacement Tips, O.C. White
Pens Washable Markers
These handy pens allow for very accurate flux dispensing on the work area, while eliminating dripping and flooding. Very easily cleaned and maintained with alcohol or water, depending on flux use.
  • Reusable flux applicator, tips and cartridge
  • Low cost, 7 cc capacity
  • Fine point and flexible shafts
  • Controlled application/no mess
Used with non-wash, wash, or strong acid flux and excellent for spot soldering on IC chips.