Model GM 80CR HEPA Filter Vacuum, Nilfisk
Supplier: Nilfisk-Advance
01790150
76239-528EA 3413.79 USD
76239-528
Model GM 80CR HEPA Filter Vacuum, Nilfisk
This GM 80CR vacuum cleaner is specifically designed and packaged for use in cleanrooms. It features unique four-stage filtration system, by which it safely collects and retains fine dust and powders.
  • Equipped with an ULPA exhaust filter (captures 99.999% of ultra fine particles, down to and including 0.12 µm)
  • Motor is interference suppressed and will not affect electronic equipment in cleanrooms
  • Disposable paper bag with 2.25 gal. capacity captures the bulk of collected debris
  • Conductive plastic hose provides ESD protection
  • Wheeled trolley adds mobility and glides over all surfaces
  • Positive twist safety latches offer security by preventing the container from opening if the vacuum is dropped
Features extra-large main filter that ensures a steady, even airflow which extends filter life and eliminates premature clogging. The microfilter, a final step in pre-filtration, protects the motor and acts as a barrier to even bacteria-size particles. Motor thermal protection device prevents overheating caused by failure to keep filters clean or by accidental blockage in a nozzle or hose.
The vacuum cleaner and accessories are packaged in cleanroom compatible polyethylene bags to prevent contamination from packing materials during shipping.
Certifications: Meets cleanroom standards up to and including Class 10 (ISO 4).