Sodium thiocyanate solution, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Sodium rhodanide

Management of Change (MOC) category = NI

J.T. Baker®
BAKR3939-09 BAKR3939-09
JT3939-09CS 16292.01 USD
CAJT3939-09 JT3939-09
Sodium thiocyanate solution, BAKER ANALYZED®, J.T.Baker®
Sodium thiocyanate
Formula: NaSCN
MW: 81.07 g/mol
MDL Number: MFCD00011123
CAS Number: 540-72-7

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Density (g/mL) at 20°C 1.3064 - 1.3122
Insoluble Matter ≤0.002 %
Carbonate (as Na₂CO₃) ≤0.1 %
Chloride (Cl) ≤0.002 %
Sulfate (SO₄) ≤0.005 %
Sulfide (S) ≤0.0005 %
Ammonium (NH₄) ≤0.001 %
Reducing Agents (x10⁻⁵ meq/gm), For Information Only
Heavy Metals (as Pb) ≤2 ppm
Trace Impurities - Iron (Fe) ≤1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR