Zinc stearate 12.5 - 14.0 % RSN, Hyqual®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Stearic acid zinc saltZinc(II) stearate

Management of Change (MOC) category = N

MACR4644-32 MACR4644-32 MACR4644-32
MK4644-32CS 1315.13 USD
MK4644-32 CAMK4644-32 JT4644-32
Zinc stearate 12.5 - 14.0 % RSN, Hyqual®, Macron Fine Chemicals™
Zinc stearate
Formula: C₃₆H₇₀O₄Zn
MW: 632.34 g/mol
Density: 1.10 g/cm³
Flash Pt: 279.00 °C
MDL Number: MFCD00013031
CAS Number: 557-05-1
https://us.vwr.com/stibo/web/std.lang.all/25/73/2302573.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (Zn(C₁₈H₃₅O₂)₂) 12.5 - 14.0 %
Appearance Passes Test
Identification A Passes Test
Identification B ≥54 °C
Arsenic (As) ≤1.500 ppm
Loss on Drying ≤1.25 %
Lead (Pb) ≤0.001 %
Alkalies and Alkaline Earths (as SO₄) ≤1.0 %
Thru U.S. No. 325 Sieve ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR