D-Ring Non-Stretch Low Charging Cable Tie, Desco Industries
Supplier: Desco Industries
35656
76243-596EA 11.47 USD
76243-596
D-Ring Non-Stretch Low Charging Cable Tie, Desco Industries
Ties
D-Ring fastening is ideal for bundling trays and IC tubes.
  • Adjustable, releasable and reusable - easy to install
  • Straps should be tested regularly to make sure they retain their ESD properties
  • Operating temperature: 300 °F max
  • Melting temperature: 459 °F