Polyester Labels, Brady
32464
76241-564EA 176.2 USD
76241-564
Polyester Labels, Brady
These labels feature permanent polyester (B-619) which is suitable for marking components in control panels, electronic components and circuit boards, faceplates and electrical receptacles.
  • Leaves no adhesive residue when removed, so it's ideal for permanent or temporary applications
  • Matte finish material with permanent acrylic adhesive can be thermal transfer printed or dot matrix printed
  • Good smudge and solvent resistance in temperatures of –70 to 145 °C (–94 to 293 °F)
Abrasion-, smear-, corrosion-, humidity-, UV- and weather-resistant.
Applications include circuit board ID - after process, circuit board ID - through hole top, component and equipment ID, general and industrial ID and panel labeling.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted.