Hot Air Stations, JBC
Supplier: JBC
JTSE-1QA JT-1B JT-1QD JTSE-1A
76251-442EA 10684 USD
76251-442 MSPP-JTSE-1QA 76245-696 76247-696
Hot Air Stations, JBC
Soldering Supplies Soldering Stations
The JT is a high powered hot air station to repair all types of SMD quickly and safely , even the biggest QFPs and PLCCs.
  • Using the menu to personalize over 20 parameters to help manage the soldering process
The unit features two work modes -manual or profile- to manage efficiently the temperature and the hot air flow.
The station works with the JT-TA heater and incorporates a USB connector (type B).
JT-1QD for even the biggest QFPs and PLCCs. Closed-loop control with external thermocouple provides precise temperature control of the component/PCB, regardless of the distance to the heater. It helps to protect the components or a specific area on the PCB. Type K thermocouple: Vacuum: 30% / 228 mmHg.
A safety measure which ensures the heat is automatically cut off when the tool is in the JT-SE stand.
Delivery information: JT-1QD comes with a complete extractor desk and a heater stand with the quick nozzle changer.