Dichloromethane ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED® for biotechnology, J.T.Baker®

Warning

Synonyms: Methane dichlorideMethylene chloride

Management of Change (MOC) category = N

BAKR9348-98 BAKR9348-99 BAKR9348-98 BAKR9348-50 BAKR9348-50 BAKR9348-99
JT9348-98CS 1249.03 USD
JT9348-98 JT9348-99 CAJT9348-98 CAJT9348-99 JT9348-50 CAJT9348-50
Dichloromethane ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED® for biotechnology, J.T.Baker®
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Density: 1.32 g/cm³
MDL Number: MFCD00000881
CAS Number: 75-09-2
UN: 1593
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (by GC, corrected for water, exclusive of preservative) ≥99.8 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤233
Residue after Evaporation ≤2.0 ppm
Titrable Acid (meq/g) ≤0.0003
Chloride (Cl) ≤10 ppm
Water (by KF, coulometric) ≤0.010 %
Preservative (cyclohexene) ≥50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR