Infrared Heaters, TPI Corporation
CH-2212-1C
76245-304EA 938.28 USD
76245-304
Infrared Heaters, TPI Corporation
CH 2212-1C has two 1000 W elements that can be switched on / off by toggle switches to produce 1000 or 2000 W of heat.
  • Operates with a 20 A molded plug (not included)
  • 60° symmetrical heat pattern
Delivery information: Quartz tubes and built in wire guards included.
Caution: Interior or protected outdoor use only.