Tartan™ Tapes, 3M
Supplier: 3M
7010373410
76243-134CS 379.68 USD
76243-134
Tartan™ Tapes, 3M
Tapes
Tartan™ Tapes are general purpose packing tape ideal for non-critical box sealing applications.
  • High edge tear and split resistance
  • Polypropylene backing with synthetic rubber adhesive
  • Easy unwind and smooth dispensing
The polypropylene film backing is easy to unwind and resists abrasion, moisture and scuffing. The backing easily conforms around edges and on rough surfaces for a tight seal.
Resists abrasion, moisture, chemicals and scuffing for long-lasting performance. Excellent adhesion to recycled liner paper and fiberboard.