Bacitracin, potency min. 60 Units/mg

Warning

AC22610-0050
103511-648EA 149 USD
103511-648
Bacitracin, potency min. 60 Units/mg
Bacitracin
Formula: C₆₆H₁₀₃N₁₇O₁₆S₁
Melting Pt: 221 °C - 225 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062640
CAS Number: 1405-87-4
EINECS: 215-786-2

Order Now

Specification Test Results

Appearance White to off-white to beige Powder
Infrared spectrum Conforms
Loss on drying ≤5.0 % (1 g, 60 °C, 3 h) (P2O5) (not exceeding 0.1 kPa)
Sulfated ash ≤1.0 %
Potency ≥60 IU/mg (on dried substance)
Related substances (Eur. Pharm. (HPLC)) Passes test for composition

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR