Microcentrifuge Tubes with Screw Caps, Polypropylene, BIO PLAS INC.
Supplier: Bio Plas
4204SLS 4203SLS 4200SB 4200SLS 4202SLS 4215RSL 4202SL 4204SL 4203SL
60828-812CS 2838.47 USD
60828-812 60828-814 60828-816 47734-878 60828-808 60828-818 60828-820 60828-822 60828-810
Microcentrifuge Tubes with Screw Caps, Polypropylene, BIO PLAS INC.
Tubes Microcentrifuge Tubes
Siliconized screw cap microcentrifuge tubes are designed for packaging, transporting and storage of any diagnostic test, especially in the area of biotechnology, biological products and reagents.
  • Positive screw cap seal with an O-Ring provides safe, reliable secure fit for repeated cap closures
  • Withstands centrifugation at 25000 ×g
  • Available as both sterile and non sterile
Features extremely low surface adhesion which is ideal for protein or nucleic acid studies to prevent any possible loss of sample due to adherence to the inner walls of the tube allowing complete sample recovery. A tight seal closure eliminates accidental opening during incubation, spinning or transporting.
Ordering information: Screw cap can be ordered separately.