Magnesium carbonate 40.0 - 43.5%, powder USP, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Magnesium Carbonate

Management of Change (MOC) category = R

MACR5942-32 MACR5942-32
MK594232EA 748.58 USD
MK594232 CAMK594232
Magnesium carbonate 40.0 - 43.5%, powder USP, FCC, Macron Fine Chemicals™
Magnesium carbonate
Formula: CMgO₃
MW: 84.31 g/mol
Density: 3.03 g/cm³ (16 °C)
MDL Number: MFCD00064632
CAS Number: 546-93-0
EINECS: 208-915-9

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (MgO) 40.0 - 43.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - Microbiological Passes Test
USP - Soluble Salts ≤ 1.0 %
USP - Acid-Insoluble Substances ≤ 0.05 %
USP - Arsenic (As) ≤ 3 ppm
USP - Limit of Calcium (Ca) ≤ 0.45 %
USP - Iron (Fe) ≤ 0.02 %
FCC - Assay (MgO) 40.0 - 43.5 %
FCC - Identification Passes Test
FCC - Soluble Salts ≤ 1.0 %
FCC - Acid-Insoluble Substances ≤ 0.05 %
FCC - Lead (Pb) ≤ 2 mg/kg
FCC - Calcium Oxide ≤ 0.6 %
Appearance (bulky, white powder) Passes Test
Solubility in Dilute Sulfuric Acid Passes Test
Residue on US 325 Mesh ≤ 0.3 %
Tapped Density ≥ 0.43 g/ml
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR