Dual Conductor Metal Wrist Straps are durable, premium performance metal wristbands that create a reliable electrical contact to the user’s wrist.
- Comfortable, designed not to pinch or pull hair
- Durable metal band is long-wearing and easy to clean
- 3.4 mm plug size
- Includes 5' coil cord, with one megohm current limiting resistor located at socket end
- Made in the United States of America
Wristbands offer superior contact with skin for reliable removal of generated static charges. The dual-wire conductor ensures redundancy, so that even if one dual-wire wrist strap wire is severed, the operator has reliable path-to-ground with other wire. These wristbands provides 360 degree protection in multiple size options to allow for conformance to most user’s wrists.
Certifications: Meets the resistance requirements of ANSI/ESD S20.20-2014, EIA 62, and ANSI/ESD S1.1.
Ordering information: Use wrist straps with SCS continuous monitors (sold separately).