Fluor-A-Pure™ PFA Tubing, Thermoplastic Biologic (TBL)
Supplier: Thermoplastic Biologic (TBL)
TBL-PFA-250X375-20 TBL-PFA-250X375-50
76268-356EA 916.74 USD
76268-356 76239-714
Fluor-A-Pure™ PFA Tubing, Thermoplastic Biologic (TBL)
Tubing Transfer Tubing
Fluor-A-Pure™ has been specifically developed for the most extreme chemical and ultra-high purity service conditions.
  • Manufactured from 100% virgin-grade high-performace resins
  • Chemically inert to most industrial chemicals and solvents
  • Low permeability and animal-derived-ingredient free
  • Higher thermal stability than FEP
It has excellent flexibility compared to other grades of PFA tube and is ideal for use with flaring tools and compression fittings.
The tubing has superior electrical insulation, non-sticky, and non flammable.
Flammability: UL 94 VO Rated (Resists combustion and spread of flames).
Certifications: In compliance with CFR Title 21 Section 177.1550, meets or exceeds ASTM D 6867-03, U.S. Pharmacopoeia Class VI, Cytotoxicity criteria.