Dual Conductor Metal Wrist Straps are durable, premium performance metal wristbands that create a reliable electrical contact to the user’s wrist.
- Comfortable design will not pinch or pull hair
- Durable metal band is long wearing and easy to clean
- Superior contact with skin for reliable removal of generated static charges
- 3.4 mm plug size
- Available in three sizes
- Easy to use; no adjustment necessary
- Made in the United States of America
The wristband provides 360 degree protection. The dual wire conductor provides redundancy, so that even if one dual-wire wrist strap wire is severed, the operator has a reliable path-to-ground with the other wire.
Certifications: Meets the resistance requirements of ANSI/ESD S20.20-2014, EIA 625.
Ordering information: Use wristbands with SCS dual-wire coil cords and SCS continuous monitors (sold separately).