Mouse H-2Db, PE (Phycoerythrin)
Supplier: MBL International
TB-5009-1
103258-514EA 2307.69 USD
103258-514
Mouse H-2Db, PE (Phycoerythrin)
Proteins and Peptides
MHC tetramers are used for the detection of antigen-specific T-cell populations.
  • Class I
  • gp276-286 specific
MHC-peptide tetramer complexes recognize antigen specific T-cells at the single cell level, allowing for accurate measurements of the functional immune response in cancer, infectious diseases, and autoimmune diseases. Due to their increased avidity, MHC tetramers stably bind to cognate TcRs and are evaluated using flow cytometry.
Used in immunology, immune monitoring.