Mouse Serum Non-Sterile, Rockland Immunochemicals
D208-00-0100
75934-936EA 1179.43 USD
75934-936
Mouse Serum Non-Sterile, Rockland Immunochemicals
Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Mouse Serum is used as a component of bioassays, immunoassays or enzyme assays. Mouse Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Mouse Serum is also available as a sterile preparation for use as a cell culture supplement. Mouse Serum is ideal for investigators in Cancer, Immunology and Cell Biology research. Mouse Serum can be used as a blocking agent to treat plastic surfaces, membrane or tissue after they have been sensitized with primary antibody or antigen. It provides an alternative to fish gel and other blocking agents. It is effective in reducing nonspecific binding of proteins to reaction surfaces, thereby maximizing signal-to-noise ratio. This blocking agent is recommended for use in immunoassays where the primary antibody was produced in mouse, as a source of non-specific serum protein or on tissue for immunohistochemical applications.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal