Avidin, Cy2®
Supplier: Rockland Immunochemicals
A003-11
10800-282EA 198.48 USD
10800-282
Avidin, Cy2®
Proteins and Peptides
Avidin Cy2 Conjugated
  • electrophoretically purified avidin isolated from egg white
  • conjugated to the chromophore Cy2
  • purified chromatographically
Avidin Cy2 Conjugated is suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
Avidin Cy2 Conjugated is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting.