Would you like to log in again?
Nuclear Pore Complex Antibody, Rockland Immunochemicals
Supplier: Rockland Immunochemical
900-301-D98
75930-580
Nuclear Pore Complex Antibody, Rockland Immunochemicals
Mouse Anti-Nuclear Pore Complex Antibody
  • Titrated value sufficient to run approximately 10 mini blots.
  • Useful in Immunofluorescence
Nuclear Pore Complex Antibody detects Nuclear Pore Complex proteins. The nuclear pore complex (NPC) is a very large structure made up of at least 50 different proteins that span the double membrane of the nuclear envelope functioning as a gateway for macromolecular traffic between the cytoplasm and the nucleus. Discrete nuclear pore complex proteins or nucleoporins such as NUP98, NUP180 and p62 have been implicated in autoimmune disease and cancer. Patients with primary biliary cirrhosis (PBC) frequently produce autoantibodies against p62 and NUP180 while NUP98 translocations have been found in patients with acute myelogenous leukemia (AML). Anti-Nuclear Pore Complex Antibody is ideal for investigators involved in Cancer and Immunology Research.
Anti-Nuclear Pore Complex Antibody is suitable for use in IF. Specific conditions for reactivity should be optimized by the end user
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty