Would you like to log in again?
Protein G Antibody Peroxidase Conjugated, Rockland Immunochemicals
Supplier: Rockland Immunochemical
200403138-0100
76235-086
Protein G Antibody Peroxidase Conjugated, Rockland Immunochemicals
Rabbit Anti-Protein G Peroxidase Conjugated Antibody
  • Purified IgG Fraction
  • HRP
  • ELISA
Protein G is a protein that has the property of binging to immunoglobulins. It is a 65-kDa cell surface protein that is commonly used for purifying antibodies through binding to the Fab and Fc regions.
Protein G was originally isolated from Streptococcal bacteria. It is similar in properties to Protein A but has unique IgG binding specificities. Native protein G also binds albumin, however Rockland uses recombinant forms of Protein G that only bind to immunoglobulins. Horseradish Peroxidase (HRP) is an enzyme that utilize organic peroxide compounds as electron donors. Naturally provides protection for plants against pathogens, but can be utilized in molecular biology to convert various substrates to detectable compounds (such as in Western Blotting and ELISAs). Anti-Protein G Antibody is ideal for researchers in Immunology, Cancer, and Microbiology.
Anti-Protein G Peroxidase Conjugated Antibody has been assayed against 1.0 ug of Protein G [Streptococcus species] in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:20,000 to 1:100,000 of the reconstitution concentration is suggested for this product.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty