Would you like to log in again?
Protein G Antibody Fluorescein Conjugated, Rockland Immunochemicals
Supplier: Rockland Immunochemical
200402138-0100
76235-072EA 370.25 USD InStock
76235-072
Protein G Antibody Fluorescein Conjugated, Rockland Immunochemicals
Antibodies
Rabbit Anti-Protein G Fluorescein Conjugated Antibody
  • Purified IgG Fraction
  • FITC
  • multiplex, immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting
Protein G is a protein that has the property of binging to immunoglobulins. It is a 65-kDa cell surface protein that is commonly used for purifying antibodies through binding to the Fab and Fc regions.
Protein G was originally isolated from Streptococcal bacteria. It is similar in properties to Protein A but has unique IgG binding specificities. Native protein G also binds albumin, however Rockland uses recombinant forms of Protein G that only bind to immunoglobulins.
FITC Conjugated Anti-Protein G antibody is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty