Would you like to log in again?
Luciferase Antibody Biotin Conjugated, Rockland Immunochemicals
Supplier: Rockland Immunochemical
200106150-0100
76234-888
Luciferase Antibody Biotin Conjugated, Rockland Immunochemicals
Goat Anti-Luciferase Antibody Biotin Conjugated
  • Purified IgG Fraction
  • Biotin
  • ELISA
Anti luciferase Antibody recognizes luciferase that is commonly used in biological research as a reporter to assess the transcriptional activity in cells that are transfected with a genetic construct containing the luciferase gene under the control of a promoter of interest. Luciferase can also be used to detect the level of cellular ATP in cell viability assays or for kinase activity assays. Additionally proluminescent molecules that are converted to luciferin upon activity of a particular enzyme can be used to detect enzyme activity in coupled or two-step luciferase assays. Such substrates have been used to detect caspase activity and cytochrome P450 activity, among others.
Anti-Luciferase Peroxidase conjugated antibody is produced from Photinus pyralis (Firefly) which produces a green light with a wavelength of 562 nm. Expect a band at approximately 60.7 kDa in size corresponding to Luciferase by western blotting in the appropriate cell lysate or extract. Anti-Luciferase has been assayed against 1.0 ug of Luciferase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:80,000 to 1:310,000 of the reconstitution concentration is suggested for this product.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty