Would you like to log in again?
MONKEY IgG, Rockland Immunochemicals
Supplier: Rockland Immunochemical
017-0102-0001
10823-674EA 148.85 USD InStock
10823-674
MONKEY IgG, Rockland Immunochemicals
Antibodies
MONKEY IgG Whole Molecule
  • control or standard reagent
  • SDS, Western Blotting, and ELISA experiments
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The whole IgG molecule possesses both the F(c) region, recognized by high-afinity Fc receptor proteins, as well as the F(ab) region possessing the epitope-recognition site. Both heavy and light chains of the antibody molecule are present.
Monkey IgG whole molecule can be utilized as a control or standard reagent in SDS, Western Blotting, and ELISA experiments.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty