Swine IgG Fab
Supplier: Rockland Immunochemicals
014-0105-0001
75932-418EA 241.55 USD
75932-418
Swine IgG Fab
Antibodies
Swine IgG Fab Fragment
  • control or standard reagent
  • SDS, Western Blotting, and ELISA experiments
Swine IgG Fab Fragment - secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The F(ab) fragment is the portion of the antibody that binds to the antigen target. The immunoglobulin Fab also possesses one constant and one variable region of both the heavy and light chain.
Swine IgG Fab fragment reagents are ideal for ELISA, western blotting, Immunohistochemistry, as well as other antibody detection methods.