Would you like to log in again?
Swine IgG F(ab')2, Rockland Immunochemicals
Supplier: Rockland Immunochemical
014-0104-0001
75932-416EA 214.97 USD InStock
75932-416
Swine IgG F(ab')2, Rockland Immunochemicals
Antibodies
Swine IgG F(ab')2 Fragment
  • control or standard reagent
  • SDS, Western Blotting, and ELISA experiments
Swine IgG F(ab')2 Fragment is a proteolytic fragment of immunoglobulin G (IgG) obtained by limited digestion with the enzyme pepsin under controlled conditions of temperature, time and pH. Swine IgG F(ab')2 molecules lack the Fc portion of Swine IgG and therefore receptors that bind Swine IgG Fc will not bind Swine IgG F(ab')2 molecules. This product possesses the F(ab')2 fragment, recognized by the two F(ab) fragments yielded from the digestion of the antibody below the disulfide bond hinge region.
Swine IgG F(ab')2 Fragment reagents are ideal for ELISA, western blotting, Immunohistochemistry, as well as other antibody detection methods.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty