Would you like to log in again?
rRat CCL7 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
012001V76-0010 012001V76-0100 012001V76-1000 012001V76-0002
75933-910 75930-052 75933-904 75930-058
rRat CCL7 Protein, Rockland Immunochemicals
Recombinant Rat CCL7 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Monocyte Chemotactic Protein 3 (MCP-3), also called CCL7, is chemokine produced by macrophages and some tumor cell lines. MCP-3 signals through three different G protein-coupled receptors, CCR1, CCR2, and CCR3. CCL7 chemoattracts monocytes and can regulate macrophage function. Recombinant rat MCP-3 is a non-glycosylated protein, containing 75 amino acids, with a molecular weight of 8.6 kDa.
Monocyte Chemotactic Protein-3 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Monocyte Chemotactic Protein-3 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty